Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perilipin-3, C-terminus Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABS482MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS482MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS482MI Supplier MilliporeSigma Supplier No. ABS482

Rabbit Polyclonal Antibody

This Anti-Perilipin-3 antibody, C-terminus is validated for use in western blotting and immunocytochemistry for the detection of Perilipin-3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Perilipin-3
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS without preservatives with 10% glycerol.
Gene Symbols Plin3; M6prbp1; Tip47
Host Species Rabbit
Immunogen Linear peptide corresponding to the C-terminus of mouse Perilipin-3, C-terminus.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_080112
Target Species Mouse
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.