Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Periostin Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABT253MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT253MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT253MI Supplier MilliporeSigma Supplier No. ABT253
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Periostin Antibody (C-term) is an antibody against Periostin (C-term) for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Periostin
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.
Gene Symbols Postn; OSF-2
Host Species Rabbit
Immunogen A 188-amino acid bacterial fusion protein from the C-terminal region of mouse periostin.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001185694
Target Species Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.