Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peripherin-2 Mouse anti-Mouse, Human, Rat, Clone: 6B10.1, MilliporeSigma™

Catalog No. MABN293MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN293MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN293MI Supplier MilliporeSigma Supplier No. MABN293
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Peripherin-2 Antibody, clone 6B10.1 is a highly specific mouse monoclonal antibody, that targets Peripherin and has been tested in western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peripherin-2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6B10.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000313
Gene Symbols PRPH2, PRPH, RDS, TSPAN22
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the cytoplasmic domain of human Peripherin-2.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.