Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peripherin Rabbit anti-Human, Rat, Mouse, Bovine, Pig, Polyclonal, MilliporeSigma™

Catalog No. AB1530MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1530MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1530MI Supplier MilliporeSigma Supplier No. AB1530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Peripherin Antibody is validated for use in immunohistochemistry, immunohistochemistry (paraffin), western blotting for the detection of Peripherin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peripherin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal antisera, no preservatives.
Gene Symbols PRPH; Peripherin; PRPH1; peripherin; NEF4
Host Species Rabbit
Immunogen Electrophoretically pure trp-E-peripherin fusion protein [Dev. Brain Res. (1990) 57:235-248], containing all but the 4 N terminal amino acids of rat peripherin. Fusion protein purified from bacterial inclusion bodies by DEAE-cellulose chromatography in 6
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_006253
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.