Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peripherin Mouse anti-Bovine, Human, Mouse, Pig, Rat, Clone: 8G2, MilliporeSigma™

Catalog No. MAB1527MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
300 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1527MI 300 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1527MI Supplier MilliporeSigma Supplier No. MAB1527
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Peripherin Antibody, clone 8G2 is an antibody against Peripherin for use in immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peripherin
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8G2
Conjugate Unconjugated
Formulation UnPurified mouse culture supernatant containing no preservatives.
Gene Accession No. NM_006262.3
Gene Symbols PRPH, PRPH1, Peripherin., peripherin, NEF4
Host Species Mouse
Immunogen Electrophoretically pure trp-E-peripherin fusion protein [Dev. Brain Res. (1990). 57:235-248] containing all but the 4 N-terminal amino acids of rat peripherin. Fusion protein purified from bacterial inclusion bodies by DEAE-cellulose chromatography in 6M
Purification Method Unpurified
Quantity 300 μL
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.