Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peroxiredoxin-2 Rabbit anti-Mouse, Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABN1011MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1011MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1011MI Supplier MilliporeSigma Supplier No. ABN1011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Peroxiredoxin-2 antibody is an antibody against Peroxiredoxin-2 for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peroxiredoxin-2
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum without preservatives.
Gene Symbols Prdx2; Tdpx1; Tpx
Host Species Rabbit
Immunogen Ovalbumin-conjugated linear peptide corresponding to the C-terminus of mouse Peroxiredoxin-2.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_035693
Target Species Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.