Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PET112-like (yeast), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004233 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-233 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-233 Supplier Abnova Corporation Supplier No. H00005188A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PET112L.

Sequence: SAAKQVFEELWKREGKTPGQIVSEKQLELMQDQGALEQLCHSVMEAHPQVVMDVKNRNPRAINKLIGLVRKATQSRADPVMIKEILEKKLS

Specifications

Antigen PET112-like (yeast)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PET112L.
Formulation 50% glycerol
Gene PET112L
Gene Accession No. NM_004564
Gene Alias HSPC199/PET112
Gene Symbols PET112L
Host Species Mouse
Immunogen PET112L (NP_004555, 466 a.a. ∼ 556 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5188
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.