Learn More
PEX19, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA128131
Description
This gene is necessary for early peroxisomal biogenesis. It acts both as a cytosolic chaperone and as an import receptor for peroxisomal membrane proteins (PMPs). Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs) are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. The peroxisomal biogenesis disorders are a heterogeneous group with at least 14 complementation groups and with more than 1 phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle. Defects in this gene are a cause Zellweger syndrome (ZWS).
PA1-28131 detects PEX19 from mouse, rat, human samples.PA1-28131 has been successfully used in Western blot applications.The PA1-28131 immunogen is: Synthetic peptide, corresponding to amino acids 1-299 of PEX19.
Specifications
PEX19 | |
Polyclonal | |
PEX19 | |
D1S2223E, FLJ55296, HK33, PMP1, PMPI, PXF, PXMP1 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
5824 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
Unconjugated | |
P40855, Q8VCI5, Q9QYU1 | |
PEX19 | |
Synthetic peptide, corresponding to amino acids 1-299 of PEX19. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.