Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGD2 Synthase/PTGDS Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

Supplier:  Novus Biologicals NB11059909

 View more versions of this product

Catalog No. NB11059909


Only null left
Add to Cart

Description

Description

PGD2 Synthase/PTGDS Polyclonal specifically detects PGD2 Synthase/PTGDS in Human, Mouse, Rat, Porcine, Bovine, Equine, Feline, Fish, Primate, Rabbit samples. It is validated for Western Blot, Simple Western.
Specifications

Specifications

PGD2 Synthase/PTGDS
Polyclonal
Unconjugated
PBS and 30% Glycerol with 0.1% Sodium Azide
Beta-trace protein, cerebrin-28, EC 5.3.99.2, Glutathione-independent PGD synthase, glutathione-independent PGD synthetase, lipocalin-type prostaglandin D synthase, Lipocalin-type prostaglandin-D synthase, L-PGDS, PDS, PGD2, PGD2 synthase, PGDS2, PGDSLPGDS, prostaglandin D synthase, prostaglandin D2 synthase (21kD, brain), prostaglandin D2 synthase 21kDa (brain), Prostaglandin-D2 synthase, prostaglandin-H2 D-isomerase
Rabbit
21 kDa
0.1 mL
Mast Cell Markers, Neuroscience, Signal Transduction
5730
Human, Mouse, Rat, Pig, Bovine, Equine, Feline, Fish, Primate, Rabbit
IgG
Western Blot
1 mg/mL
Western Blot 2 ug/ml, Simple Western 1:50
B2R727
PTGDS
The epitope recognized by this antibody maps to region within residue 25-100 of human Prostaglandin D Synthase/Lipocalin. [Swiss-Prot# P41222]
Affinity Purified
RUO
Primary
Human, primate, mouse, rat, bovine, porcine, feline, equine and canine. Immunogen sequence has 87% homology to sheep and 82% homology to rabbit.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only