Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phosphoglycerate kinase 2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004241 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-241 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-241 Supplier Abnova Corporation Supplier No. H00005232A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PGK2.

The PGK2 gene encodes a testis-specific form of phosphoglycerate kinase (EC 2.7.2.3), which catalyzes the reversible conversion of 1,3-diphosphoglycerate to 3-phosphoglycerate during glycolysis, generating one molecule of ATP. See also PGK1 (MIM 311800), which is ubiquitously expressed in all somatic tissues and maps to chromosome Xq13.[supplied by OMIM

Sequence: DIMAKAQKNGVRITFPVDFVTGDKFDENAQVGKATVASGISPGWMGLDCGPESNKNHAQVVAQARLIVWNGP

Specifications

Antigen phosphoglycerate kinase 2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PGK2.
Formulation 50% glycerol
Gene PGK2
Gene Accession No. NM_138733
Gene Alias PGK-2/PGKB/PGKPS/dJ417L20.2
Gene Symbols PGK2
Host Species Mouse
Immunogen PGK2 (NP_620061, 268 a.a. ∼ 339 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5232
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.