Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UCH-L1/PGP9.5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | UCH-L1/PGP9.5 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.1-0.5 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224719SS
![]() |
Novus Biologicals
NBP224719SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP224719
![]() |
Novus Biologicals
NBP224719 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
UCH-L1/PGP9.5 Polyclonal specifically detects UCH-L1/PGP9.5 in Human, Primate, Monkey samples. It is validated for Western Blot.Specifications
UCH-L1/PGP9.5 | |
Western Blot 0.1-0.5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Primate, Monkey | |
P09936 | |
7345 | |
A portion of amino acids 100-150 of human UCH-L1/PGP9.5 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1 | |
UCHL1 | |
IgG | |
Protein G purified | |
The amino acid sequence used as immunogen is 100% homologous in human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title