Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHC1, Mouse, Clone: 3G1, Abnova™

Catalog No. 89021392 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-392 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-392 Supplier Abnova Corporation Supplier No. H00001911M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant PHC1.

This gene is a homolog of the Drosophila polyhomeotic gene, which is a member of the Polycomb group of genes. The gene product is a component of a multimeric protein complex that contains EDR2 and the vertebrate Polycomb protein BMH1. The gene product, the EDR2 protein, and the Drosophila polyhomeotic protein share 2 highly conserved domains, named homology domains I and II. These domains are involved in protein-protein interactions and may mediate heterodimerization of the protein encoded by this gene and the EDR2 protein. [provided by RefSeq

Sequence: PVGCSQLLKESEKPLQTGLPTGLTENQSGGPLGVDSPSAELDKKANLLKCEYCGKYAPAEQFRGSKRFCSMTCAKRYNVSCSHQFRLKRKKMKEFQEANY*

Specifications

Antigen PHC1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant PHC1.
Formulation PBS with no preservative; pH 7.4
Gene PHC1
Gene Accession No. NM_004426
Gene Alias EDR1/HPH1/RAE28
Gene Symbols PHC1
Host Species Mouse
Immunogen PHC1 (NP_004417, 751 a.a. ∼ 851 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1911
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.