Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHD finger protein 8 Rabbit anti-Human, Mouse, Rat, Chimpanzee, Bovine, Rabbit, Rhesus Macaque, Canine, Polyclonal, MilliporeSigma™

Catalog No. 09868MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-868-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-868-MI Supplier MilliporeSigma Supplier No. 09868

Rabbit Polyclonal Antibody

Anti-PHD finger protein 8 Antibody is a Rabbit Polyclonal Antibody for detection of PHD finger protein 8 also known as Histone lysine demethylase PHF8, PHD finger protein 8 and has been validated in western blot, immunocytochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHD finger protein 8
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols PHF8; ZNF422
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human PHD finger protein 8.
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001171825
Target Species Bovine, Canine, Chimpanzee, Human, Mouse, Rabbit, Rat, Rhesus Monkey
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.