Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHD finger protein 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017949 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-949 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-949 Supplier Abnova Corporation Supplier No. H00005252D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human PHF1 protein.

This gene encodes a Polycomb group protein. The protein is a component of a histone H3 lysine-27 (H3K27)-specific methyltransferase complex, and functions in transcriptional repression of homeotic genes. The protein is also recruited to double-strand breaks, and reduced protein levels results in X-ray sensitivity and increased homologous recombination. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MAQPPRLSRSGASSLWDPASPAPTSGPRPRLWEGQDVLARWTDGLLYLGTIKKVDSAREVCLVQFEDDSQFLVLWKDISPAALPGEELLCCVCRSETVVPGNRLVSCEKCRHAYHQDCHVPRAPAPGEGEGTSWVCRQCVFAIATKRGGALKKGPYARAMLGMKLSLPYGLKGLDWDAGHLSNRQQSYCYCGGPGEWNLKMLQCRSCLQWFHEACTQCLSKPLLYGDRFYEFECCVCRGGPEKVRRLQLRWVDVAHLVLYHLSVCCKKKYFDFDREILPFTSENWDSLLLGELSDTPKGERSSKLLSALNSHKDRFISGREIKKRKCLFGLHARMPPPVEPPTGDGALTSFPSGQGPGGGVSRPLGKRRRPEPEPLRRRQKGKVEELGPPSAVRNQPEPQEQRERAHLQRALQASVSPPSPSPNQSYQGSSGYNFRPTDARCLPSSPIRMFASFHPSASTAGTSGDSGPPDRSPLELHIGFPTDIPKSAPHSMTASSSSVSSPSPGLPRRSAPPSPLCRSLSPGTGGGVRGGVGYLSRGDPVRVLARRVRPDGSVQYLVEWGGGGIF

Specifications

Antigen PHD finger protein 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human PHF1 protein.
Formulation PBS with no preservative; pH 7.4
Gene PHF1
Gene Accession No. NM_024165.1
Gene Alias MTF2L2/PCL1/PHF2
Gene Symbols PHF1
Host Species Rabbit
Immunogen PHF1 (NP_077084.1, 1 a.a. ∼ 567 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 5252
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.