Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF17, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017170 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-170 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-170 Supplier Abnova Corporation Supplier No. H00079960B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PHF17 protein.

Sequence: MKRGRLPSSSEDSDDNGSLSTTWSQNSRSQHRRSSCSRHEDRKPSEVFRTDLITAMKLHDSYQLNPDEYYVLADPWRQEWEKGVQVPVSPGTIPQPVARVVSEEKSLMFIRPKKYIVSSGSEPPELGYVDIRTLADSVCRYDLNDMDAAWLELTNEEFKEMGMPELDEYTMERVLEEFEQRCYDNMNHAIETEEGLGIEYDEDVVCDVCQSPDGEDGNEMVFCDKCNICVHQACYGILKVPEGSWLCRTCALGVQPKCLLCPKKGGAMKPTRSGTKWVHVSCALWIPEVSIGSPEKMEPITKVSHIPSSRWALVCSLCNEKFGASIQCSVKNCRTAFHVTCAFDRGLEMKTILAENDEVKFKSYCPKHSSHRKPEESLGKGAAQENGAPECSPRNPLEPFASLEQNREEAHRVSVRKQKLQQLEDEFYTFVNLLDVARALRLPEEVVDFLYQYWKLKRKVNFNKPLITPKKDEEDNLAKREQDVLFRRLQLFTHLRQDLERVMIDTDTL

Specifications

Antigen PHF17
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PHF17 protein.
Formulation PBS with no preservative; pH 7.4
Gene PHF17
Gene Accession No. NM_024900
Gene Alias FLJ22479/JADE1/KIAA1807
Gene Symbols PHF17
Host Species Mouse
Immunogen PHF17 (NP_079176.2, 1 a.a. ∼ 509 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79960
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.