Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF19 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE992MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE992MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE992MI Supplier MilliporeSigma Supplier No. ABE992

Rabbit Polyclonal Antibody

Anti-PHF19 Antibody is a highly specific rabbit polyclonal antibody, that targets PHD finger protein 19 and has been tested in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF19
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with 0.01% thimerosal and 0.1% BSA.
Gene Symbols HTA2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human PHF19.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001009936
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.