Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHD finger protein 2, Mouse, Clone: 3E7, Abnova™

Catalog No. 89001476 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-476 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-476 Supplier Abnova Corporation Supplier No. H00005253M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant PHF2.

This gene encodes a protein which contains a zinc finger-like PHD (plant homeodomain) finger, distinct from other classes of zinc finger motifs, and a hydrophobic and highly conserved domain. The PHD finger shows the typical Cys4-His-Cys3 arrangement. PHD finger genes are thought to belong to a diverse group of transcriptional regulators possibly affecting eukaryotic gene expression by influencing chromatin structure. [provided by RefSeq

Sequence: ATVPVYCVCRLPYDVTRFMIECDACKDWFHGSCVGVEEEEAPDIDIYHCPNCEKTHGKSTLKKKRTWHKHGPGQAPDVKPVQNGSQLFIKELRSRTFPS

Specifications

Antigen PHD finger protein 2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant PHF2.
Formulation ascites with no preservative
Gene PHF2
Gene Accession No. NM_005392
Gene Alias GRC5/JHDM1E/KIAA0662/MGC176680
Gene Symbols PHF2
Host Species Mouse
Immunogen PHF2 (NP_005383, 2 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5253
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.