Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

integrator complex subunit 12, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007967 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-967 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-967 Supplier Abnova Corporation Supplier No. H00057117B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PHF22 protein.

INTS12 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1; MIM 180680) and U2 (RNU2; MIM 180690) (Baillat et al., 2005 [PubMed 16239144]).[supplied by OMIM

Sequence: MAATVNLELDPIFLKALGFLHSKSKDSAEKLKALLDESLARGIDSSYRPSQKDVEPPKISSTKNISIKQEPKISSSLPSGNNNGKVLTTEKVKKEAEKRPADKMKSDITEGVDIPKKPRLEKPETQSSPITVQSSKDLPMADLSSFEETSADDFAMEMGLACVVCRQMMVASGNQLVECQECHNLYHRDCHKPQVTDKEANDPRLVWYCARCTRQMKRMAQKTQKPPQKPAPAVVSVTPAVKDPLVKKPETKLKQETTFLAFKRTEVKTSTVISGNSSSASVSSSVTSGLTGWAAFAAKTSSAGPSTAKLSSTTQNNTGKPATSSANQKPVGLTGLATSSKGGIGSKIGSNNSTTPTVPLKPPPPLTLGKTGLSRSVSCDNVSKVGLPSPSSLVPGSSSQLSGNGNSGTSGPSGSTTSKTTSESSSSPSASLKGPTSQESQLNAMKRLQMVKKKAAQKKLKK

Specifications

Antigen integrator complex subunit 12
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PHF22 protein.
Formulation No additive
Gene INTS12
Gene Accession No. NM_020395.2
Gene Alias INT12/PHF22/SBBI22
Gene Symbols INTS12
Host Species Mouse
Immunogen PHF22 (NP_065128.2, 1 a.a. ∼ 462 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 57117
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.