Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phosphorylase kinase, gamma 1 (muscle), Mouse, Clone: 2B4, Abnova™

Catalog No. 89000878 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-878 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-878 Supplier Abnova Corporation Supplier No. H00005260M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant PHKG1.

This gene is a member of the Ser/Thr protein kinase family and encodes a protein with one protein kinase domain and two calmodulin-binding domains. This protein is the catalytic member of a 16 subunit protein kinase complex which contains equimolar ratios of 4 subunit types. The complex is a crucial glycogenolytic regulatory enzyme. This gene has two pseudogenes at chromosome 7q11.21 and one at chromosome 11p11.12. [provided by RefSeq

Sequence: MTRDEALPDSHSAQDFYENYEPKEILGRGVSSVVRRCIHKPTSQEYAVKVIDVTGGGSFSPEEVRELREATLKEVDILRKVSGHPNIIQLKDTYETNTFFFLVFDLMKRGELFDYLTEKVTLSEKETRKIMRALLEVICTLHKLNIVHRDLKPENILLDDNMNIKLTDFGFSCQLEPGERLREVCGTPSYLAPEIIECSMNEDHPGYGKEVDMWSTGVIMYTLLAGSPPFWHRKQMLMLRMIMSGNYQFGSPEWDDYSDTVKDLVSRFLVVQPQNRYTAEEALAHPFFQQYLVEEVRHFSPRGKFKVIALTVLASVRIYYQYRRVKPVTREIVIRDPYALRPLRRLIDAYAFRIYGHWVKKGQQQNRAALFENTPKAVLLSLAEEDY

Specifications

Antigen phosphorylase kinase, gamma 1 (muscle)
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 2B4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant PHKG1.
Formulation PBS with no preservative; pH 7.4
Gene PHKG1
Gene Accession No. BC051327
Gene Alias PHKG
Gene Symbols PHKG1
Host Species Mouse
Immunogen PHKG1 (AAH51327, 1 a.a. ∼ 387 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5260
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.