Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHLPP1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514994
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
PHLPP1 is a protein phosphatase that specifically mediates dephosphorylation of ′Ser-473′ of AKT1, a protein that regulates the balance between cell survival and apoptosis through a cascade that primarily alters the function of transcription factors that regulate pro- and antiapoptotic genes.
Specifications
PHLPP1 | |
Polyclonal | |
PHLPP1 | |
PH domain leucine-rich repeat-containing protein phosphatase; PH domain leucine-rich repeat protein phosphatase; EC 3.1.3.16; Pleckstrin homology domain-containing family E protein 1; Suprachiasmatic nucleus circadian oscillatory protein; hSCOP; PHLPP1; PHLPP; KIAA0606; PLEKHE1; SCOP | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
23239 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
O60346 | |
PHLPP1 | |
KLH conjugated synthetic peptide between 531-560 amino acids from the N-terminal region of human PHLPP1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction