Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphatidylserine Mouse anti-Vertebrates, AlexaFluor™488, Clone: 1H6, MilliporeSigma™

Catalog No. 16256MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-256-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-256-MI Supplier MilliporeSigma Supplier No. 16256
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Phosphatidylserine using this Anti-Phosphatidylserine Antibody, clone 1H6, Alexa Fluor 488 validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphatidylserine
Applications Flow Cytometry
Classification Monoclonal
Clone 1H6
Conjugate Alexa Fluor 488
Formulation Purified mouse monoclonal IgG in buffer containing PBS containing 1% BSA, 0.05% Tween, 0.05% sodium azide. Liquid at 4ºC.
Host Species Mouse
Immunogen Liposomes containing 70% phosphatidylserine and 30% phosphatidylglycerol.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage Stable for 1 year at from date of receipt.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.