Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphatidylserine Mouse anti-Vertebrates, Clone: 1H6, MilliporeSigma™

Catalog No. 05719MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-719-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-719-MI Supplier MilliporeSigma Supplier No. 05719
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Phosphatidylserine Antibody, clone 1H6 is an antibody against Phosphatidylserine for use in flow cytometry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphatidylserine
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 1H6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG in buffer containing 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol. Liquid at −20°C.
Host Species Mouse
Immunogen Liposomes containing 70% phosphatidylserine and 30% phosphatidylglycerol
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage Stable for 1 year at from date of receipt.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.