Learn More
Phospho-Abl (Tyr245) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-Abl (Tyr245) |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Gene Symbols | ABL1; JTK7; p150; bcr/abl; v-abl; c-ABL; ABL |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated peptide corresponding to amino acid region encompassing human c-Abl Tyr245. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.