Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Acetyl CoA Carboxylase (Ser79) Rabbit anti-Human, Mouse, Rat, Canine, Chicken, Bovine, Polyclonal, MilliporeSigma™

Catalog No. 07303MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-303-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-303-MI Supplier MilliporeSigma Supplier No. 07303
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho-Acetyl CoA Carboxylase (Ser79) Antibody is an antibody against phospho-Acetyl CoA Carboxylase (Ser79) for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Acetyl CoA Carboxylase (Ser79)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A purified in buffer containing in 0.1M Tris-Glycine (pH7.4), 150mM NaCl, with 0.05% NaN3.
Gene Symbols ACACA; ACC-alpha; ACCA; ACC; ACC1; ACAC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide (C-HM RSSM[pS]GLHLVK) corresponding to amino acid 73-85 of rat Acetyl CoA Carboxylase.
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_942131
Target Species Bovine, Canine, Chicken, Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.