Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Akt (Ser473) Mouse anti-Human, Mouse, Rat, Vertebrates, Clone: 6F5, MilliporeSigma™

Catalog No. 051003MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-100-3MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-100-3MI Supplier MilliporeSigma Supplier No. 051003
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-Akt (Ser473) Antibody, clone 6F5 detects level of phospho-Akt (Ser473) and has been published and validated for use in ELISA, western blot, flow cytometry, immunofluoresence, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Akt (Ser473)
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6F5
Conjugate Unconjugated
Formulation Purified mouse monolconal IgG1κ in 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05%NaN3
Gene Accession No. NP_001014431
Gene Symbols AKT, C-AKT, EC 2.7.11.1, MGC99656, MGC99656, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA, RAC-PK-alpha
Host Species Mouse
Immunogen Peptide encompassing and including phosphorylated Ser473 of human Akt1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Vertebrates
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.