Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Akt (Thr308) Rabbit anti-Human, Mouse, Rabbit, Bovine, Chicken, Clone: NL50, MilliporeSigma™

Catalog No. 05802RMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-802-RMI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-802-RMI Supplier MilliporeSigma Supplier No. 05802R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Detect phospho-Akt (Thr308) using this Anti-phospho-Akt (Thr308) Antibody, clone 50-1C-25 validated for use in western blot, immunofluoresence, immunocytochemistry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Akt (Thr308)
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone NL50
Conjugate Unconjugated
Formulation Protein A purified recombinant Rabbit Monoclonal supernatant in 0.1M Tris-Glycine (pH7.4) 150mM NaCl, and 0.05% NaN3 with 40% glycerol.
Gene Accession No. NP_001014431
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide containing the Thr308 sequence and surrounding amino acids of human, mouse, rat, bovine and chicken Akt1.
Purification Method Protein A purfied
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) AKT, C-AKT, EC 2.7.11.1, MGC99656, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA, RAC-PK-alpha
Target Species Bovine, Chicken, Human, Mouse, Rabbit
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.