Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Akt (Thr308) Rabbit anti-Human, Mouse, Rat, Bovine, Xenopus, Canine, Polyclonal, MilliporeSigma™

Catalog No. 071398MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-139-8MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-139-8MI Supplier MilliporeSigma Supplier No. 071398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-phospho-Akt (Thr308) Antibody is validated for use in immunohistochemistry (paraffin) for the detection of phospho-Akt (Thr308).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Akt (Thr308)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Antigen affinity purified rabbit serum in 0.1M Tris-Glycine (pH7.4), 150mM NaCl, with 0.05% NaN3.
Gene Symbols AKT ; C-AKT; EC 2.7.11.1; MGC99656; PKB ; PKB-ALPHA ; RAC; RAC-ALPHA ; RAC-PK-alpha
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide containing the Thr308 sequence and surrounding amino acids of human, mouse, rat, bovine and chicken Akt1. Human and mouse Akt2 shares 11/12 amino acids and rat Akt3 shares 10/12 amino acids with the peptide immunogen seque
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001014431
Target Species Bovine, Canine, Human, Mouse, Rat, Xenopus species
Content And Storage 2-8°C for 12 months
Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.