Learn More
MilliporeSigma™ anti-Phospho-Akt (Thr308), Polyclonal
Supplier: MilliporeSigma™ 071398/DEL

Description
Akt/PKB is a Ser/Thr kinase and a major known effecter of the PI3 Kinase pathway. It is involved in multiple signaling pathways that relate to many biological processes including metabolism, apoptosis, cell cycle control, angiogenesis, differentiation, and cell growth and proliferation. In mammals, three isoforms of Akt (Akt1/PKCα, Akt2/PKBβ, and Akt3/PKBγ) exists. They exhibit a high degree of homology, but differ slightly in the localization of their regulatory phosphorylation sites. Akt1 is the predominant isoform that is in most tissues and is thought to have a dominant role in growth, survival, embryonic development, and adipocyte differentiation. Akt2 is correlated with the regulation of glucose homeostasis and is the predominant isoform expressed in insulin-responsive tissues. Akt3 is abundant in brain tissue. Each Akt isoform is composed of three functionally distinct regions: an N-terminal Pleckstrin Homology (PH) domain that provides a lipid-binding module, a central catalytic domain containing Thr308, and a C-terminal hydrophobic motif containing Ser473. The activation of AKT is dependent on a dual regulatory mechanism that requires both its translocation to the plasma membrane and dual phosphorylation on Thr308 and Ser473 by PDK1 and the TORC2 complex, respectively.
Host Species:Rabbit
Clone:
Isotype:IgG
Species Reactivity:Human, Mouse, Rat, Bovine, enopus, Canine
Epitope:Thr308 (Catalytic Domain)
Immunogen:KLH-conjugated synthetic peptide containing the Thr308 sequence and surrounding amino acids of human, mouse, rat, bovine and chicken Akt1. Human and mouse Akt2 shares 11/12 amino acids and rat Akt3 shares 10/12 amino acids with the peptide immunogen sequence.
Molecular Weight:60kDa
Immunohistochemistry (Paraffin)

Specifications
Phospho-Akt (Thr308) | |
0.5mg/mL | |
Rabbit | |
100 μL | |
Cell Signaling | |
Bovine, Canine, Human, Mouse, Rat, Xenopus species |
Polyclonal | |
Unconjugated | |
Affinity Purified | |
RUO | |
Primary | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.