Learn More
Description
Specifications
Specifications
| Antigen | phospho-AKT (Thr34) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 100μL of epitope specific affinity purified rabbit polyclonal IgG in 50% affinity storage buffer (PBS containing 1.0mg/mL BSA and 0.05% sodium azide) and 50% glycerol. |
| Gene Symbols | AKT1; RAC; PRKBA; MGC99656; RAC-ALPHA; RAC-PK-alpha; C-AKT; PKB; AKT |
| Host Species | Rabbit |
| Immunogen | KLH conjugated peptide corresponding to amino acid region encompassing the human phospho-AKT (Thr34); Sequence highly conserved in mouse and rat. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.