Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Akt1/PKBα (Ser473) Mouse anti-Canine, Human, Mouse, Rat, AlexaFluor™488, Clone: 11E6, MilliporeSigma™

Catalog No. 16305MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-305-MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-305-MI Supplier MilliporeSigma Supplier No. 16305
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect phospho-Akt1/PKBα (Ser473) using this Anti-Akt1 Antibody, clone 11E6, Alexa Fluor 488 validated for use in flow cytometry, immunocytochemistry, immunofluorescence and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Akt1/PKBα (Ser473)
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 11E6
Conjugate Alexa Fluor 488
Gene Accession No. NM_001014431.1; NM_001014432.1; NM_005163.2
Gene Symbols AKT1, RAC, PRKBA, MGC99656, RAC-ALPHA, RAC-PK-alpha, C-AKT, PKB, AKT
Host Species Mouse
Immunogen KLH conjugated synthetic peptide containing a pSer that corresponds to amino acid residues around phosphoserine 473.
Purification Method Purified from serum-free cell culture supernatant by subsequent thiophilic adsorption and size exclusion chromatography
Quantity 50 μg
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage 4°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.