Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Akt1/PKBα (Ser473) Rabbit anti-Human, Mouse, Clone: SK703, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 05736
Description
Detect phospho-Akt1/PKBα (Ser473) using this Anti-Akt1 Antibody, clone SK703 validated for use in western blotting.
Specifications
phospho-Akt1/PKBα (Ser473) | |
Monoclonal | |
Unconjugated | |
NM_005163 | |
Peptide containing a pSer that corresponds to amino acid position 473 of human Akt1/PKBa | |
100 μg | |
AKT1, RAC, PRKBA, RAC-ALPHA, PKB, MGC99656, RAC-PK-alpha, C-AKT, AKT | |
2-8°C in undiluted aliquots for up to 12 months | |
IgG |
Western Blot | |
SK703 | |
Protein A purified antibody in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide. | |
Rabbit | |
Protein A purfied | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction