Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Akt1/PKBα (Thr308) Rabbit anti-Human, Mouse, Clone: NL50, MilliporeSigma™

Catalog No. 05802MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-802-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-802-MI Supplier MilliporeSigma Supplier No. 05802

Rabbit Monoclonal Antibody

Anti-Akt1 Antibody, clone NL50 is an antibody against phospho-Akt1/PKBα (Thr308) for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Akt1/PKBα (Thr308)
Applications Western Blot
Classification Monoclonal
Clone NL50
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative.
Gene Accession No. NM_005163
Host Species Rabbit
Immunogen peptide containing the sequence MK[pT]FC in which pT corresponds to phospho-threonine at residue 308 of mouse Akt
Purification Method Protein A purfied
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) AKT1, RAC, PRKBA, RAC-ALPHA, PKB, MGC99656, RAC-PK-alpha, C-AKT, AKT
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.