Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-AMPK α (Thr172) Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07681MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-681-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-681-MI Supplier MilliporeSigma Supplier No. 07681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho-AMPK α (Thr172) Antibody is an antibody against phospho-AMPK α (Thr172) for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-AMPK α (Thr172)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A Purified rabbit IgG in a solution of 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.135M NaCl, and 0.035% sodium azide as a preservative.
Gene Symbols PRKAA1; MGC57364; AMPK; MGC33776; AMPKa1; AMPK1
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids surrounding phosphorylated Thr172 of human AMP-activated protein kinase alpha 2 (AMPKα2)
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_006251.5; NM_206907.3
Target Species Human, Rat
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.