Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-AMPKα (Thr172) Rabbit anti-Human, Rat, Mouse, Horse, Chicken, Bovine, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07626
Description
Anti-phospho-AMPKα (Thr172) Antibody is an antibody against phospho-AMPKα (Thr172) for use in western blotting.
Specifications
phospho-AMPKα (Thr172) | |
Polyclonal | |
Purified rabbit polyclonal serum in buffer containing 0.1M Tris-glycine, pH 7.4, 150mM NaCl and 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Bovine, Chicken, Equine, Human, Mouse, Rat | |
IgG |
Western Blot | |
Unconjugated | |
PRKAA1; MGC57364; AMPK; AMPK1; MGC33776; AMPKa1 | |
KLH-conjugated, synthetic peptide corresponding to amino acids surrounding phosphorylated Thr172 of human AMP-activated protein kinase alpha 2 (AMPKα2). The immunogenic site is conserved in AMPKα1, isoform 1 (Thr174), AMPKα1, isoform 2 | |
100 μg | |
NP_996790 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction