Learn More
Phospho-Androgen Receptor (Ser81) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-Androgen Receptor (Ser81) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. |
| Gene Symbols | AR; androgen receptor; RP11-383C12.1; AIS; DHTR; HUMARA; KD; NR3C4; SBMA; SMAX1; TFM |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated synthetic peptide corresponding to amino acids including and surrounding Ser81 of human Androgen Receptor. |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.