Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Androgen Receptor (Ser81) Rabbit anti-Human, MilliporeSigma™

Catalog No. 04078MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-078-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-078-MI Supplier MilliporeSigma Supplier No. 04078
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-phospho-Androgen Receptor (Ser81) Antibody detects level of phospho-Androgen Receptor (Ser81) and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Androgen Receptor (Ser81)
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation 100μl of cell culture supernatant with 0.1% sodium azide
Gene Accession No. NM_000044.2, NM_001011645.1
Host Species Rabbit
Immunogen A synthetic phospho-peptide surrounding Ser81 of human Androgen Receptor.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) AR, AIS, DHTR, SMAX1, NR3C4, TFM, OTTHUMP00000061928, KD, HUMARA, SBMA
Target Species Human
Content And Storage −20°C for 24 months
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.