Learn More
Phospho-ATM (Ser1981) Mouse anti-Human, Mouse, Clone: 10H11.E12, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-ATM (Ser1981) |
| Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 10H11.E12 |
| Conjugate | Unconjugated |
| Formulation | Protein G purified mouse IgG in 0.014Mphosphate buffer, pH 7.6, with 0.175MNaCl, 0.07 % Sodium Azide and 30% glycerol. Liquid at −20°C. |
| Gene Accession No. | NP_000042 |
| Gene Symbols | ATM, AT, TELO1, ATC, DKFZp781A0353, MGC74674, ATD, ATA, AT1, T-PLL, ATE, TEL1, ATDC, TPLL |
| Host Species | Mouse |
| Immunogen | KLH-conjugated, synthetic peptide corresponding to amino acids 1974-1988 (SLAFEEG[pS]QSTTISS) of human ATM. The immunizing sequence has 11/12 identical amino acids in mouse and rat. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.