Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Bad (Ser75) Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB10423
Description
Anti-Phospho-Bad (Ser75) Antibody detects level of Phospho-Bad (Ser75) and has been published and validated for use in western blotting.
Specifications
| Phospho- Bad (Ser75) | |
| Polyclonal | |
| Purified immunoglobulins in PBS with 0.09% NaN3. | |
| Rabbit | |
| Protein G followed by IAP | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| BBC2; BBC6; BCL2L8 | |
| This antibody is generated in rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding pS75 of human Bad. | |
| 100 μg | |
| NP_116784 | |
| 2-8°C in undiluted aliquotes for up to 6 months, −20°C for long term. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction