Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Bad (Ser99) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB10424MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10424MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10424MI Supplier MilliporeSigma Supplier No. AB10424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Phospho-Bad (Ser99) Antibody is validated for use in western blotting for the detection of Phospho-Bad (Ser99).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Bad (Ser99)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified in PBS with 0.09% NaN3.
Gene Symbols BBC2; BBC6; BCL2L8
Host Species Rabbit
Immunogen This antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S99 of human Bad.
Purification Method Protein G followed by IAP
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_116784
Target Species Human
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months, −20°C for long term. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.