Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-BMK1/Erk5 (Thr218/Tyr220) Rabbit anti-Human, Mouse, Rat, Bovine, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07507
Description
This Anti-phospho-BMK1/Erk5 (Thr218/Tyr220) Antibody is validated for use in western blotting for the detection of phospho-BMK1/Erk5 (Thr218/Tyr220).
Specifications
phospho-BMK1/Erk5 (Thr218/Tyr220) | |
Polyclonal | |
Affinity purified immunoglobulin in 0.1M Tris-Glycine (pH 7.4), 150mM NaCl, with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Bovine, Human, Mouse, Rat | |
IgG |
Inhibition Assays, Western Blot | |
Unconjugated | |
MAPK7; ERK5; BMK1; ERK-5; ERK4; PRKM7 | |
KLH-conjugated, synthetic peptide corresponding to amino acids 213-224 of human BMK1/Erk5 | |
200 μL | |
NM_139033 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction