Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-phospho-BRCA1 (Ser1497), Polyclonal,

Catalog No. 07007MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-007-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-007-MI Supplier MilliporeSigma™ Supplier No. 07007

Specifications

Antigen phospho-BRCA1 (Ser1497)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation of 0.1M Tris glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of 30% glycerol
Gene Symbols BRCA1; RNF53; BRCAI; PSCP; BRCC1; IRIS
Host Species Rabbit
Immunogen KLH-conjugated, synthetic phospho-peptide corresponding to residues 1489-1500 of human BRCA1 (C-KEPGVERS[pS]PSK)
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_007294
Target Species Human
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.