Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-phospho-c-Jun (Ser63), Polyclonal,

Catalog No. 06828MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-828-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-828-MI Supplier MilliporeSigma™ Supplier No. 06828

Specifications

Antigen phospho-c-Jun (Ser63)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Tris, 0.05% sodium azide, 5mg/ml BSA before the addition of glycerol to 30%
Gene Symbols JUN; p39; AP1; c-Jun
Host Species Rabbit
Immunogen BSA conjugated synthetic peptide (C-RAKNSDLLT[pS]PDV) corresponding to amino acids 54-66 of human c-Jun
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_010591
Target Species Human, Mouse, Rat
Content And Storage −20°C for 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.