Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Caldesmon (Ser789) Rabbit anti-Canine, Ferret, Pig, Polyclonal, MilliporeSigma™

Catalog No. 07156MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-156-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-156-MI Supplier MilliporeSigma Supplier No. 07156

Rabbit Polyclonal Antibody

Detect phospho-Caldesmon (Ser789) using this Anti-phospho-Caldesmon Antibody validated for use in EA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Caldesmon (Ser789)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Symbols CALD1; Caldesmon; CAD; CDM; L-CAD; MGC21352; NAG22; H-CAD
Host Species Rabbit
Immunogen peptide corresponding to amino acids 782-793 (C-QSVDKVT[pS]PTKV) of human Caldesmon
Purification Method Protein A chromatography
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_033140
Target Species Canine, Ferret, Pig
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.