Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-CaM Kinase II Mouse anti-Rat, Clone: 22B1, MilliporeSigma™

Catalog No. 05533MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-533-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-533-MI Supplier MilliporeSigma Supplier No. 05533
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect phospho-CaM Kinase II using this Anti-phospho-CaM Kinase II Antibody, α subunit, (Thr286), clone 22B1 validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-CaM Kinase II, α subunit, (Thr286)
Applications Western Blot
Classification Monoclonal
Clone 22B1
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_015981 and others
Gene Symbols CAMK2A, KIAA0968, CAMKA
Host Species Mouse
Immunogen Peptide corresponding to residuues 281-294 of rat Cam Kinase II alpha-subunit; recognizes phosphorylated CaM Kinase II on Threonine 286
Purification Method Protein G chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.