Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-CREB (Ser133) Mouse anti-Human, Mouse, Rat, Canine, Clone: 10E9, MilliporeSigma™

Catalog No. 05667MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-667-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-667-MI Supplier MilliporeSigma Supplier No. 05667
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-CREB (Ser133) Antibody, clone 10E9 is a Mouse Monoclonal Antibody for detection of phospho-CREB (Ser133) also known as active transcription factor CREB and has been validated in western blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-CREB (Ser133)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 10E9.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ from serum-free cell culture supernatant, lyophilized from 1 mL of PBS, 0.09% sodium azide, PEG, and sucrose. Lyophilized.
Gene Accession No. NM_134442
Gene Symbols CREB1, CREB, MGC9284
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide (REILSRRP(pS)YR) corresponding to amino acids 125-135 of human CREB. The immunizing sequence is identical in human and mouse. Clone 10E9.
Purification Method Thiophilic adsoption and size exclusion chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.