Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-phospho-Cyclin E1 (Thr77), Clone: EPR2242(2), Rabbit Monoclonal,
Supplier: MilliporeSigma™ MABE582
Description

Specifications
phospho-Cyclin E1 (Thr77) | |
Monoclonal | |
Unconjugated | |
Rabbit | |
Unpurified | |
Primary | |
Human | |
IgG |
Immunohistochemistry, Western Blot | |
EPR2242(2) | |
Rabbit Monoclonal in buffer containing PBS with up to 0.1% sodium azide, 0.05% BSA, and 50% glycerol. | |
Synthetic peptide corresponding to human phospho-Cyclin E1 (Thr77). | |
100 μL | |
CCNE1, CCNE | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction