Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-EGFR (Thr669) Mouse anti-Human, Clone: 5F10, MilliporeSigma™

Catalog No. 04281MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-281-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-281-MI Supplier MilliporeSigma Supplier No. 04281
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-EGFR (Thr669) Antibody, clone 5F10 detects level of phospho-EGFR (Thr669) and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-EGFR (Thr669)
Applications Western Blot
Classification Monoclonal
Clone 5F10
Conjugate Unconjugated
Formulation 100μg of mouse monoclonal IgG1 liquid in 0.01mg/mL PBS containing 0.09% sodium azide, PEG, and sucrose with 50 % glycerol.
Gene Accession No. NM_005228.3; NM_201282.1; NM_201283.1; NM_201284.1
Gene Symbols EGFR, ERBB1, ERBB, mENA
Host Species Mouse
Immunogen KLH-conjugated synthetic peptide encompassing the surrounding amino acids of Thr 669 in human EGFR.
Purification Method Thiophilic adsoption and size exclusion chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.