Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-EGFR (Tyr1069) Rabbit anti-Human, Mouse, Pig, Primate, Horse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 09310MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-310-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-310-MI Supplier MilliporeSigma Supplier No. 09310

Rabbit Polyclonal Antibody

This Anti-phospho-EGFR (Tyr1069) Antibody is validated for use in ELISA, immunocytochemistry, western blotting for the detection of phospho-EGFR (Tyr1069).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-EGFR (Tyr1069)
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Antigen affinity purified rabbit polyclonal in 0.1M Tris-Glycine (pH 7.4) 150mM NaCl, with 0.05% sodium azide.
Gene Symbols ERBB; ERBB1; HER1 ; PIG61; mENA ; EC 2.7.10.1
Host Species Rabbit
Immunogen Tyrosine-phosphorylated epidermal growth factor receptor.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_005228.3
Target Species Equine, Human, Mouse, Pig, Primate, Rat
Content And Storage 2-8°C for up to 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.