Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-EGFR (Tyr1173) Mouse anti-Human, Mouse, AlexaFluor™488, Clone: 9H2, MilliporeSigma™

Catalog No. 16244MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-244-MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-244-MI Supplier MilliporeSigma Supplier No. 16244
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-EGFR (Tyr1173) Antibody, clone 9H2, Alexa Fluor 488 detects level of phospho-EGFR (Tyr1173) and has been published and validated for use in FC and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-EGFR (Tyr1173)
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 9H2
Conjugate Alexa Fluor 488
Gene Accession No. NM_005228.3; NM_201282.1; NM_201283.1; NM_201284.1
Gene Symbols EGFR, ERBB1, ERBB, mENA
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide containing the Y1173 motif (...NAE[pY1173]LRV...) of the human EGFR
Quantity 25 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 4°C for one year
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.