Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-eNOS/NOS III (Ser114) Rabbit anti-Bovine, Human, Polyclonal, MilliporeSigma™

Catalog No. 07357MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-357-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-357-MI Supplier MilliporeSigma Supplier No. 07357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect phospho-eNOS/NOS III (Ser114) using this Anti-phospho-eNOS/NOS III (Ser114) Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-eNOS/NOS III (Ser116)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.2M Tris-glycine, pH 7.4, 0.15M NaCl, 0.1mM EDTA, and 0.05% sodium azide before the addition of glycerol to 30%
Gene Symbols NOS3; cNOS; ECNOS; eNOS; EC-NOS; NOSIII
Host Species Rabbit
Immunogen peptide corresponding to amino acids 106-118 of human eNOS/NOS III (Ser116) (PRKLQGRPpSPGPP-C)
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000603
Target Species Bovine, Human
Content And Storage −20°C for 12 months.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.